Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PP14B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562861
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562861 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562861 Supplier Invitrogen™ Supplier No. PA562861
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ELEIDVDELL DMESDDARAA RVKELLV Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.

This gene is thought to play a role in a complex developmental pathway and the regulation of genes involved in cerebellar development. Mutations in this gene have been associated with autosomal recessive spinocerebellar ataxia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PP14B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PPP1R14B
Gene Accession No. Q96C90
Gene Alias AOM172; Phi1; PHI-1; Phosphatase holoenzyme inhibitor 1; phospholipase C beta-3 neighboring gene protein; phospholipase c neighboring; phospholipase C-beta-3 neighbouring gene protein; PLCB3N; PNG; Ppp1r14b; protein phosphatase 1 regulatory inhibitor subunit 14B; protein phosphatase 1 regulatory subunit 14B; protein phosphatase 1, regulatory (inhibitor) 5 subunit 14B; protein phosphatase 1, regulatory (inhibitor) subunit 14B; SOM172; ubiquitous PKC-potentiated PP1 inhibitor
Gene Symbols PPP1R14B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PP14B. Recombinant protein control fragment (Product #RP-101495).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26472
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.