Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PP14B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5104037
Description
Antibody detects endogenous levels of total PPP1R14B.
This gene is thought to play a role in a complex developmental pathway and the regulation of genes involved in cerebellar development. Mutations in this gene have been associated with autosomal recessive spinocerebellar ataxia.
Specifications
PP14B | |
Polyclonal | |
Unconjugated | |
PPP1R14B | |
AOM172; Phi1; PHI-1; Phosphatase holoenzyme inhibitor 1; phospholipase C beta-3 neighboring gene protein; phospholipase c neighboring; phospholipase C-beta-3 neighbouring gene protein; PLCB3N; PNG; Ppp1r14b; protein phosphatase 1 regulatory inhibitor subunit 14B; protein phosphatase 1 regulatory subunit 14B; protein phosphatase 1, regulatory (inhibitor) 5 subunit 14B; protein phosphatase 1, regulatory (inhibitor) subunit 14B; SOM172; ubiquitous PKC-potentiated PP1 inhibitor | |
Rabbit | |
Affinity chromatography | |
RUO | |
18938, 259225, 26472 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q62084, Q8K3F3, Q96C90 | |
PPP1R14B | |
A synthesized peptide derived from human PPP1R14B(Accession Q96C90), corresponding to amino acid residues Q78-I128. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction