Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PP2A alpha Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB1002260AF350
Description
PP2A alpha Polyclonal antibody specifically detects PP2A alpha in Human, Mouse samples. It is validated for Western BlotSpecifications
PP2A alpha | |
Polyclonal | |
Western Blot | |
EC 3.1.3.16, PP2A-alpha, PP2Ac, PP2CA, PP2Calpha, protein phosphatase 2 (formerly 2A), catalytic subunit, alpha isoform, protein phosphatase 2, catalytic subunit, alpha isozyme, protein phosphatase 2A catalytic subunit, alpha isoform, Replication protein C, RP-C, serine/threonine protein phosphatase 2A, catalytic subunit, alpha isoform, serine/threonine-protein phosphatase 2A catalytic subunit alpha isoform | |
The immunogen recognized by this antibody maps to a region between residue 260 and the C-terminus (residue 309) of human Protein Phosphatase 2A using the numbering given in entry NP_004147.1 (GeneID 5516). The region is identical to that of PPP2CA (NP_00 | |
0.1 mL | |
Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Neuronal Cell Markers, Neurotransmission, Phospho Specific, Protein Phosphatase, Signal Transduction | |
5515 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction