Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PP2A alpha/beta Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA517980
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA517980 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA517980 Supplier Invitrogen™ Supplier No. PA517980
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with bovine, canine, mouse, porcine, rabbit and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.

This gene encodes the phosphatase 2A catalytic subunit. Protein phosphatase 2A is one of the four major Ser/Thr phosphatases, and it is implicated in the negative control of cell growth and division. It consists of a common heteromeric core enzyme, which is composed of a catalytic subunit and a constant regulatory subunit, that associates with a variety of regulatory subunits. This gene encodes an alpha isoform of the catalytic subunit.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PP2A alpha/beta
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene PPP2CB
Gene Accession No. P62714, P62715, P62716, P63330, P63331, P67775, P67776
Gene Alias AI115466; D8Ertd766e; fb17h07; fc05c02; fk24a01; hypothetical protein LOC550585; PP2A; Pp2a1; Pp2a2; PP2A-alpha; PP2Abeta; PP2A-beta; PP2Ac; PP2CA; PP2Calpha; PP2CB; ppp2ca; ppp2caa; PPP2CB; protein phosphatase 2 (formerly 2A), catalytic subunit, alpha isoform; protein phosphatase 2 (formerly 2A), catalytic subunit, beta isoform; protein phosphatase 2 catalytic subunit alpha; protein phosphatase 2 catalytic subunit alpha a; protein phosphatase 2 catalytic subunit beta; protein phosphatase 2, catalytic subunit, alpha isoform; protein phosphatase 2, catalytic subunit, alpha isozyme; protein phosphatase 2, catalytic subunit, beta isoform; protein phosphatase 2, catalytic subunit, beta isozyme; protein phosphatase 2,catalytic subunit alpha isozyme; protein phosphatase 2A alpha subunit; protein phosphatase 2A beta subunit; protein phosphatase 2A catalytic alpha subunit; protein phosphatase 2A catalytic subunit alpha isoform; protein phosphatase 2A catalytic subunit beta isoform; protein phosphatase 2A catalytic subunit, beta isoform; protein phosphatase 2a, catalytic subunit, alpha isoform; protein phosphatase 2A, catalytic subunit, beta isoform; protein phosphatase type 2A catalytic subunit; protein phosphatase-2A-alpha; protein phosphatase-2A-beta; R75353; replication protein C; RP C; RP-C; serine/threonine protein phosphatase 2A, catalytic subunit, alpha isoform; serine/threonine protein phosphatase 2A, catalytic subunit, beta isoform; serine/threonine-protein phosphatase 2A catalytic subunit alpha isoform; Serine/threonine-protein phosphatase 2A catalytic subunit beta isoform; testicular tissue protein Li 146; wu:fb17h07; wu:fc05c02; wu:fk24a01; zgc:110641; zgc:55808; zgc:77172
Gene Symbols PPP2CA, PPP2CB
Host Species Goat
Immunogen Synthetic peptide sequence (EPHVTRRTPDYFL) corresponding to the C-terminus amino acids of PPP2CA / PPP2CB.
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19052, 19053, 24672, 24673, 397136, 397656, 5515, 5516
Target Species Human, Mouse, Rat, Pig
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.