Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PP2A B-alpha Monoclonal Antibody (2G9)
GREENER_CHOICE

Catalog No. PIMA518064
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA518064 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA518064 Supplier Invitrogen™ Supplier No. MA518064
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody reacts very strongly with rat alpha and delta isoforms, and weakly with the human beta and rabbit gamma isoforms. This antibody is predicted to react with porcine, rabbit, chicken and xenopus samples based on sequence homology.

PPP2R2A (Protein Phosphatase 2 Regulatory Subunit Balpha) is a Protein Coding gene. Diseases associated with PPP2R2A include Lung Cancer and Hepatocellular Carcinoma. Among its related pathways are Cell Cycle, Mitotic and p70S6K Signaling. Gene Ontology (GO) annotations related to this gene include protein serine/threonine phosphatase activity and protein phosphatase regulator activity. An important paralog of this gene is PPP2R2D. The B regulatory subunit might modulate substrate selectivity and catalytic activity, and also might direct the localization of the catalytic enzyme to a particular subcellular compartment. Essential for serine/threonine-protein phosphatase 2A-mediated dephosphorylation of WEE1, preventing its ubiquitin-mediated proteolysis, increasing WEE1 protein levels, and promoting the G2/M checkpoint. The product of this gene belongs to the phosphatase 2 regulatory subunit B family. Protein phosphatase 2 is one of the four major Ser/Thr phosphatases, and it is implicated in the negative control of cell growth and division. It consists of a common heteromeric core enzyme, which is composed of a catalytic subunit and a constant regulatory subunit, that associates with a variety of regulatory subunits. The B regulatory subunit might modulate substrate selectivity and catalytic activity. This gene encodes an alpha isoform of the regulatory subunit B55 subfamily. Alternatively spliced transcript variants have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PP2A B-alpha
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 2G9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene Ppp2r2a
Gene Accession No. P36876, P63151, Q6P1F6
Gene Alias PP2Ac; PP2CA; PP2Calpha; RP-C; PP2A-alpha; protein phosphatase 2; protein phosphatase 2A catalytic subunit; replication protein C; serine/threonine protein phosphatase 2A; catalytic subunit
Gene Symbols Ppp2r2a
Host Species Mouse
Immunogen Peptide from rat PP2A isoform alpha (Immunogen sequence: 398-CASGKRKKDEISVD-411).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117104, 5520, 71978
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.