Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PP2C beta Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115784
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115784 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115784 Supplier Invitrogen™ Supplier No. PA5115784
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PP2C alpha.

The protein encoded by this gene is a member of the PP2C family of Ser/Thr protein phosphatases. PP2C family members are known to be negative regulators of cell stress response pathways. This phosphatase has been shown to dephosphorylate cyclin-dependent kinases (CDKs), and thus may be involved in cell cycle control. Overexpression of this phosphatase is reported to cause cell-growth arrest or cell death.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PP2C beta
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene PPM1B
Gene Accession No. O75688
Gene Alias B55-GAMMA; hypothetical protein; IMYPNO; IMYPNO1; Methyl-PPP2CA; MGC33570; PP2A; PP2A alpha; PP2A C terminal; PP2A L9; PP2A Leucine 9; Pp2a1; PP2A-alpha; PP2Ac; PP2CA; PP2Calpha; Ppp2ca; PR52; PR55G; protein phosphatase 2 (formerly 2A), catalytic subunit, alpha isoform; protein phosphatase 2 catalytic subunit alpha; protein phosphatase 2, catalytic subunit, alpha isoform; protein phosphatase 2, catalytic subunit, alpha isozyme; protein phosphatase 2,catalytic subunit alpha isozyme; protein phosphatase 2A alpha subunit; protein phosphatase 2A catalytic subunit alpha isoform; protein phosphatase 2a, catalytic subunit, alpha isoform; protein phosphatase type 2A catalytic subunit; protein phosphatase-2A; protein phosphatase-2A-alpha; R75353; replication protein C; RP-C; serine/threonine protein phosphatase 2A, catalytic subunit, alpha isoform; serine/threonine-protein phosphatase 2A catalytic subunit alpha isoform; unnamed protein product
Gene Symbols PPM1B
Host Species Rabbit
Immunogen A synthesized peptide derived from human PPM1B(Accession O75688), corresponding to amino acid residues E412-E462.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5495
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.