Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ PP2A B subunit isoform B56-gamma, Rabbit, Unlabeled, Polyclonal,

Catalog No. ABS1606MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ABS1606MI 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS1606MI Supplier MilliporeSigma™ Supplier No. ABS1606
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Serine/threonine-protein phosphatase 2A 56kDa regulatory subunit gamma isoform (UniProt Q13362; also known as PP2A B subunit isoform B&prime-gamma, PP2A B subunit isoform B56-gamma, PP2A B subunit isoform PR61-gamma, PP2A B subunit isoform R5-gamma, Renal carcinoma antigen NY-REN-29) is encoded by the PPP2R5C (also known as KIAA0044) gene (Gene ID 5527) in human. Protein phosphatase 2 (PP2 or PP2A) is a ubiquitously expressed heterotrimeric serine/threonine protein phosphatase with broad substrate specificity, such as Raf, MEK, and AKT. PP2A is composed of a structural A and a catalytic C subunit as the dimeric core enzyme, and a regulatory B subunit. There exist two different A subunits (PPP2R1A and PPP2R1B), two different C subunits (PPP2CA and PPP2CB), and 12 different B subunits (PPP2R2B-D, PPP2R3A-C, PPP2R4, PPP2R5A-E). The enzymatic activity of a catalytic subunit is altered upon binding an A subunit even in the absence of a B subunit. The B subunits are believed to play key roles in controlling the localization and specific activity of the A/C holoenzymes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PP2A B subunit isoform B56-gamma
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Rabbit polyclonal antibody serum with 0.05% Sodium Azide.
Gene PPP2R5C, KIAA0044
Gene Accession No. Q13362
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to a C-terminal sequence of human PP2A B subunit isoform B56-gamma.
Purification Method Unpurified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_002710
Test Specificity This antiserum is also known as M880 (Okamoto, K., et al. (1996). Mol. Cell. Biol. 16(11):6593-6602). Immunogen sequence is 100% present in human PPP2R5C spliced isoform 5 and Gamma-3, with the first amino acid missing in isoform 4 and Gamma-2. PPP2R5C spliced isoform Gamma-3 (UniProt Q13362-1) and Gamma-2 (UniProt Q13362-3) were originally referred to as B'α1 and B'α2, respectively (Tehrani, M.A., et al. (1996). J. Biol. Chem. 271(9):5164-5170). The Immunogen sequence is not present in PPP2R5C isoform Gamma-1 or any of the spliced isoforms of the other 11 types of human PP2A B subunits (PPP2R2B, PPP2R2C, PPP2R2D, PPP2R3A, PPP2R3B, PPP2R3C, PPP2R4, PPP2R5A, PPP2R5B, PPP2R5D, PPP2R5E).
Target Species Human
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Serum
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.