Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PP2A beta Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA527483
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA527483 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA527483 Supplier Invitrogen™ Supplier No. PA527483
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, mouse brain. Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (100%), Xenopus laevis (100%), Rabbit (100%), Chicken (100%), Rhesus Monkey (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes the phosphatase 2A catalytic subunit. Protein phosphatase 2A is one of the four major Ser/Thr phosphatases, and it is implicated in the negative control of cell growth and division. It consists of a common heteromeric core enzyme, which is composed of a catalytic subunit and a constant regulatory subunit, that associates with a variety of regulatory subunits. This gene encodes a beta isoform of the catalytic subunit. Two transcript variants encoding the same protein have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PP2A beta
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.01% thimerosal; pH 7
Gene Ppp2cb
Gene Accession No. P62714, P62715
Gene Alias AI115466; D8Ertd766e; PP2A beta; Pp2a2; PP2Abeta; PP2A-beta; PP2Ac; PP2CB; PPP2bc; PPP2CB; protein phosphatase 2 (formerly 2A), catalytic subunit, beta isoform; protein phosphatase 2 catalytic subunit beta; protein phosphatase 2, catalytic subunit, beta isoform; protein phosphatase 2, catalytic subunit, beta isozyme; protein phosphatase 2A catalytic alpha subunit; protein phosphatase 2A catalytic subunit, beta isoform; protein phosphatase 2a, catalytic subunit, beta isoform; protein phosphatase type 2A catalytic subunit; protein phosphatase-2A-beta; serine/threonine protein phosphatase 2A, catalytic subunit, beta isoform; serine/threonine-protein phosphatase 2A catalytic subunit beta isoform; testicular tissue protein Li 146
Gene Symbols Ppp2cb
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human PP2A beta. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19053, 5516
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.