Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PP2A-delta Rabbit anti-Human, Mouse, Rat, Polyclonal, FabGennix

Catalog No. 502208643
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-8643 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-8643 Supplier Fabgennix Inc Supplier No. PP2A121AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PP2A-delta Polyclonal antibody specifically detects PP2A-delta in Human, Mouse, Rat samples. It is validated for ELISA, Immunoprecipitation, Western Blot

Specifications

Antigen PP2A-delta
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene PPP2R5D
Gene Accession No. Q14738
Gene Alias B56D; B'delta; MGC2134; MGC8949; MRD35; PP2A B subunit isoform B56-delta; PP2A B subunit isoform B'-delta; PP2A B subunit isoform PR61-delta; PP2A B subunit isoform R5-delta; PP2A, B subunit, B' delta isoform; PP2A, B subunit, B56 delta isoform; PP2A, B subunit, PR61 delta isoform; PP2A, B subunit, R5 delta isoform; PP2CB; Ppp2r5d; protein phosphatase 2 regulatory subunit B'delta; protein phosphatase 2, regulatory subunit B (B56), delta isoform; protein phosphatase 2, regulatory subunit B', delta; protein phosphatase 2, regulatory subunit B', delta isoform; Serine/threonine protein phosphatase 2A, 56 kDa regulatory subunit, delta isoform; serine/threonine-protein phosphatase 2A 56 kDa regulatory subunit delta isoform; TEG-271; testis expressed gene 271; Tex271
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 540-600 on human Serine / Threonine-Protein Phosphatase 2A B56-Delta subunit.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21770, 363193, 5528
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.