Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PP2A-epsilon Rabbit anti-Human, Mouse, Rat, Polyclonal, FabGennix
Rabbit Polyclonal Antibody
Supplier: Fabgennix Inc PP2A131AP

Description
PP2A-epsilon Polyclonal antibody specifically detects PP2A-epsilon in Human, Mouse, Rat samples. It is validated for ELISA, Immunoprecipitation, Western BlotSpecifications
PP2A-epsilon | |
Polyclonal | |
Unconjugated | |
PPP2R5E | |
4633401M22Rik; AI449017; B56 epsilon isoform; B56beta; epsilon isoform of regulatory subunit B56, protein phosphatase 2A; Kiaa4006; PP2A B subunit isoform B56-epsilon; PP2A B subunit isoform B'-epsilon; PP2A B subunit isoform PR61-epsilon; PP2A B subunit isoform R5-epsilon; PP2A, B subunit, B' epsilon; PP2A, B subunit, B56 epsilon; PP2A, B subunit, PR61 epsilon; PP2A, B subunit, R5 epsilon; Ppp2r5e; protein phosphatase 2 regulatory subunit B', epsilon; protein phosphatase 2 regulatory subunit B'epsilon; protein phosphatase 2, regulatory subunit B (B56), epsilon isoform; protein phosphatase 2, regulatory subunit B', epsilon; protein phosphatase 2, regulatory subunit B', epsilon isoform; protein phosphatase 2A subunit beta; R5 epsilon isoform; serine/threonine protein phosphatase 2A, 56 kDa regulatory subunit, epsilon; serine/threonine-protein phosphatase 2A 56 kDa regulatory subunit epsilon isoform | |
Synthetic peptide taken within amino acid region 400-467 on human serine / Threonine-Protein Phosphatase 2A 56 kDa regulatory Epsilon subunit. | |
200 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
ELISA, Immunoprecipitation, Western Blot | |
0.5-1.5 mg/mL | |
proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8 | |
Q16537, Q61151 | |
Rabbit | |
Affinity chromatography | |
RUO | |
26932, 299147, 5529 | |
-20°C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction