Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PP2A Rabbit anti-Human, Mouse, Rat, Unconjugated, Polyclonal, Active Motif™
SDP

Catalog No. 501992641
Encompass
Change view
Click to view available options
Quantity:
10 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-2641 10 μL
50-199-2640 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-2641 Supplier Active Motif Supplier No. 39193
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes the phosphatase 2A catalytic subunit. Protein phosphatase 2A is one of the four major Ser/Thr phosphatases, and it is implicated in the negative control of cell growth and division. It consists of a common heteromeric core enzyme, which is composed of a catalytic subunit and a constant regulatory subunit, that associates with a variety of regulatory subunits. This gene encodes a beta isoform of the catalytic subunit. Two transcript variants encoding the same protein have been identified for this gene.

Specifications

Antigen PP2A
Applications ChIP Assay, Western Blot
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA, 30% glycerol and 0.035% sodium azide; pH
Gene Ppp2cb
Gene Accession No. P62714, P62715, P62716
Gene Alias AI115466; D8Ertd766e; PP2A beta; Pp2a2; PP2Abeta; PP2A-beta; PP2Ac; PP2CB; Ppp2cb; protein phosphatase 2 (formerly 2A), catalytic subunit, beta isoform; protein phosphatase 2 catalytic subunit beta; protein phosphatase 2, catalytic subunit, beta isoform; protein phosphatase 2, catalytic subunit, beta isozyme; protein phosphatase 2A catalytic alpha subunit; protein phosphatase 2A catalytic subunit, beta isoform; protein phosphatase 2a, catalytic subunit, beta isoform; protein phosphatase type 2A catalytic subunit; protein phosphatase-2A-beta; serine/threonine protein phosphatase 2A, catalytic subunit, beta isoform; Serine/threonine-protein phosphatase 2A catalytic subunit beta isoform; testicular tissue protein Li 146
Gene Symbols Ppp2cb
Host Species Rabbit
Immunogen This PP2A antibody was raised against a peptide corresponding to amino acids 288-303 of human PP2A.
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19053, 24673, 5516
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.