Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PP2AC alpha/beta Mouse anti-Human, Unconjugated, Clone: 11H12, MilliporeSigma™

Catalog No. MABE1783100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE1783-100 100 μL
MABE1783-25 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABE1783-100 Supplier MilliporeSigma Supplier No. MABE1783100UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-PP2AC alpha/beta, clone 11H12, Cat. No. MABE1783, is a highly specific mouse monoclonal antibody that targets Serine/threonine-protein phosphatase 2A catalytic subunit alpha and beta isoforms and has been tested for use in Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PP2AC alpha/beta
Applications Western Blot
Classification Monoclonal
Clone 11H12
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Serine/threonine-protein phosphatase 2A catalytic subunit alpha;Serine/threonine-protein phosphatase 2A catalytic subunit beta;PP2A-alpha;PP2A-beta
Gene Symbols PPP2CA;PPP2CB
Host Species Mouse
Immunogen KLH-conjugated linear peptide corresponding to 15 amino acids from the C-terminal region of human Protein Phosphatase 2A catalytic subunit (PP2AC). The immungen sequence is conserved between alpha and beta isoforms of PP2AC.
Molecular Weight of Antigen ∽ 35 kDa observed; 35.58 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Epigenetics & Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_002706.1;NP_001009552.1
Test Specificity Clone 11H12 detects human Serine/threonine-protein phosphatase 2A catalytic subunit alpha/beta isoforms (PP2AC alpha and beta). It targets an epitope within 15 amino acids from the C-terminal region.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.