Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PP2C alpha/PPM1A Antibody (7F12) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP237480
Description
PP2C alpha/PPM1A Monoclonal specifically detects PP2C alpha/PPM1A in Human, Primate, Monkey samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.Specifications
PP2C alpha/PPM1A | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, CyTOF-ready | |
EC 3.1.3.16, FLJ42306, MGC9201, phosphatase 2C alpha, PP2CA, PP2Calpha, PP2C-ALPHA, PPPM1A, protein phosphatase 1A, protein phosphatase 1A (formerly 2C), magnesium-dependent, alpha isoform, protein phosphatase 2C alpha isoform, Protein phosphatase 2C isoform alpha, Protein phosphatase IA, protein phosphatase, Mg2+/Mn2+ dependent, 1A | |
Mouse | |
42.4 kDa | |
0.1 mL | |
Signal Transduction | |
5494 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, Flow Cytometry, ELISA, CyTOF | |
7F12 | |
Unconjugated | |
PBS with 0.05% Sodium Azide | |
PPM1A | |
Purified recombinant fragment of human PPM1A (AA: 202-382) expressed in E. Coli. | |
Affinity Purified | |
RUO | |
Primary | |
Human, Primate, Monkey | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction