Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PP2C alpha/PPM1A Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298253PCP
Description
PP2C alpha/PPM1A Polyclonal antibody specifically detects PP2C alpha/PPM1A in Mouse samples. It is validated for Western Blot, ELISA, ImmunoprecipitationSpecifications
PP2C alpha/PPM1A | |
Polyclonal | |
Western Blot, ELISA, Immunoprecipitation | |
EC 3.1.3.16, FLJ42306, MGC9201, phosphatase 2C alpha, PP2CA, PP2Calpha, PP2C-ALPHA, PPPM1A, protein phosphatase 1A, protein phosphatase 1A (formerly 2C), magnesium-dependent, alpha isoform, protein phosphatase 2C alpha isoform, Protein phosphatase 2C isoform alpha, Protein phosphatase IA, protein phosphatase, Mg2+/Mn2+ dependent, 1A | |
Produced in rabbits immunized with purified, recombinant Mouse PP2C alpha/PPM1A (Accession#: NP_032936.1; Met 1-Trp 382) | |
0.1 mL | |
Signal Transduction | |
5494 | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA, Immunoprecipitation | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction