Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PP2C beta/PPM1B Antibody (OTI3F2), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273547IR
Description
PP2C beta/PPM1B Monoclonal specifically detects PP2C beta/PPM1B in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Flow Cytometry.Specifications
PP2C beta/PPM1B | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
PPM1B | |
Full length recombinant protein of human PP2C beta/PPM1B (NP_002697) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Canine, Monkey | |
IgG1 |
Western Blot, Flow Cytometry | |
OTI3F2 | |
Western Blot, Flow Cytometry | |
EC 3.1.3.16, MGC21657, PP2C-beta, PP2C-beta-X, PP2CBmagnesium-dependent, beta isoform, protein phosphatase 1B, Protein phosphatase 2C isoform beta, protein phosphatase 2C-like protein, protein phosphatase, Mg2+/Mn2+ dependent, 1B | |
Mouse | |
Affinity Purified | |
Stem Cell Markers | |
5495.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction