Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PP2C gamma/PPM1G Antibody, Alexa Fluor™ 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299758AF405
Description
PP2C gamma/PPM1G Polyclonal specifically detects PP2C gamma/PPM1G in Human samples. It is validated for Western Blot, ELISA.Specifications
PP2C gamma/PPM1G | |
Polyclonal | |
Western Blot, ELISA | |
EC 3.1.3.16, MGC1675, MGC2870, PP2C, gamma, PP2CG, PP2C-gamma, PP2CGAMMA, PPM1C, PPP2CG, Protein phosphatase 1C, protein phosphatase 1G, protein phosphatase 1G (formerly 2C), magnesium-dependent, gamma isoform, protein phosphatase 2, catalytic subunit, gamma isoform, protein phosphatase 2C gamma isoform, Protein phosphatase 2C isoform gamma, Protein phosphatase magnesium-dependent 1 gamma, protein phosphatase, Mg2+/Mn2+ dependent, 1G | |
Produced in rabbits immunized with purified, recombinant PP2C gamma/PPM1G (O15355; Met 317-Asp 546). | |
0.1 mL | |
Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
5496 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA | |
Alexa Fluor 405 | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction