Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PP2C gamma/PPM1G Antibody (OTI1C12), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273552C
Description
PP2C gamma/PPM1G Monoclonal specifically detects PP2C gamma/PPM1G in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
PP2C gamma/PPM1G | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
PPM1G | |
Full length recombinant protein of human PP2C gamma/PPM1G (NP_817092) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Canine | |
IgG2a |
Western Blot, Immunocytochemistry, Immunofluorescence | |
OTI1C12 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
EC 3.1.3.16, MGC1675, MGC2870, PP2C, gamma, PP2CG, PP2C-gamma, PP2CGAMMA, PPM1C, PPP2CG, Protein phosphatase 1C, protein phosphatase 1G, protein phosphatase 1G (formerly 2C), magnesium-dependent, gamma isoform, protein phosphatase 2, catalytic subunit, gamma isoform, protein phosphatase 2C gamma isoform, Protein phosphatase 2C isoform gamma, Protein phosphatase magnesium-dependent 1 gamma, protein phosphatase, Mg2+/Mn2+ dependent, 1G | |
Mouse | |
Affinity Purified | |
Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
5496.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction