Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PP2C gamma/PPM1G Mouse anti-Human, Mouse, Rat, Canine, Clone: OTI1C12, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273552
Description
PP2C gamma/PPM1G Monoclonal specifically detects PP2C gamma/PPM1G in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
PP2C gamma/PPM1G | |
Monoclonal | |
LYOPH | |
Western Blot 1:500-2000, Immunocytochemistry/Immunofluorescence 1:100 | |
EC 3.1.3.16, MGC1675, MGC2870, PP2C, gamma, PP2CG, PP2C-gamma, PP2CGAMMA, PPM1C, PPP2CG, Protein phosphatase 1C, protein phosphatase 1G, protein phosphatase 1G (formerly 2C), magnesium-dependent, gamma isoform, protein phosphatase 2, catalytic subunit, gamma isoform, protein phosphatase 2C gamma isoform, Protein phosphatase 2C isoform gamma, Protein phosphatase magnesium-dependent 1 gamma, protein phosphatase, Mg2+/Mn2+ dependent, 1G | |
Mouse | |
59.1 kDa | |
100 μg | |
Primary | |
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG2a |
Western Blot, Immunocytochemistry, Immunofluorescence | |
OTI1C12 | |
Unconjugated | |
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
PPM1G | |
Full length human recombinant protein of human PPM1G (NP_817092) produced in HEK293T cell. | |
Affinity Purified | |
Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
5496 | |
Human, Mouse, Rat, Canine | |
Lyophilized |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title