Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ PP2C Isoform delta, Mouse, Unlabeled, Clone: 2F7.1,
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ MABS1189
Description
Protein phosphatase 1D (EC 3.1.3.16; UniProt O15297; also known as Wild-type p53-induced protein phosphatase 1, PP2C-delta, Protein phosphatase 2C isoform delta, Protein phosphatase magnesium-dependent 1 delta) is encoded by the PPM1D (also known as WIP1) gene (Gene ID 8493) in human. Wild-type p53-induced phosphatase 1 (WIP1) or PP2C-delta is a serine/threonine phosphatase. Unlike PP2A or PP1 enzymes, WIP1 functions as a monomer whose catalytic activity is dependents on Mg2+ or Mn2+ and is insensitive to inhibition by okadaic acid or microcystin. Casein kinase 2 (CK2) andPP2C-delta catalyze the phosphorylation and dephosphorylation, respectively, of LSD1 on Ser131 and Ser137 residues. S131/S137 phosphorylation promots LSD1 interaction with RNF168 and RNF168-dependent 53BP1 ubiquitination and subsequent recruitment to DNA damage sites. Other known PP2C-delta targets include ATM (S367, S1981), -H2AX, CHK1 (S345) and CHK2 (T68).
Specifications
PP2C Isoform delta | |
Monoclonal | |
Please refer to lot specific datasheet. | |
Purified mouse monoclonal antibody IgG1κ in buffer containing 0.1M Tris-Glycine (pH 7.4) 150mM NaCl with 0.05% Sodium Azide. | |
O15297 | |
GST-tagged recombinant human PP2C-delta internal fragment. | |
100 μL | |
Epigenetics & Nuclear Function | |
NP_003611 | |
Human | |
Purified |
Western Blot | |
2F7.1 | |
Unconjugated | |
PPM1D, WIP1 | |
Mouse | |
Protein G Purified | |
RUO | |
Primary | |
Clone 2F7.1 targets an internal epitope conserved among the two human spliced isoforms and mouse/rat PP2C-delta (Uniprot O15297, B1WCA0, Q9QZ67). | |
Stable for 1 year at 2°-8°C from date of receipt. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction