Learn More
Invitrogen™ PPM1A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA593155
Description
Positive Samples: MCF-7, HeLa, HepG2, SH-SY5Y, Mouse liver, Mouse skeletal muscle, Rat brain; Cellular Location: Cytoplasm, Lipid-anchor, Membrane, Nucleus, cytosol Immunogen sequence: ILENVEVKGE KPAKLCVIAE CGELKEGDDG GIFPKDGSGD SHPDFPEDAD IDLKDVDKIL LITEDLKNIG NTFFKSQNWE MAIKKYAEVL RYVDSSKAVI ETADRAKLQP IALSCVLNIG ACKLKMSNWQ GAIDSCLEAL ELDPSNTKAL YRRAQGWQGL KEYDQALADL KKAQGIAPED KAIQAELLKV KQKIKAQKDK EKAVYAKMFA.
PPM1A (Protein phosphatase 1A) is also named as PPPM1A and belongs to the PP2C family. It is a serine/threonine protein phosphatase that is essential for regulating cellular stress responses in eukaryotes. PPM1A plays an important role in tumor suppression and that it negatively controls many signaling pathways which are critical for trophoblast invasion. This protein distributes in both the cytoplasm and nucleus of mammalian cells. It has 2 isoforms produced by alternative splicing.
Specifications
PPM1A | |
Polyclonal | |
Unconjugated | |
PPM1A | |
2310003C21Rik; 2900017D14Rik; AI427932; AU017636; IA; MMPa-2; Mpp alpha; MPPa-1; Pp2c1; PP2CA; PP2Calpha; PP2C-alpha; PPM1A; PPPM1A; Protein phosphatase 1A; protein phosphatase 1A (formerly 2C), magnesium-dependent, alpha isoform; protein phosphatase 1A, magnesium dependent, alpha isoform; protein phosphatase 2C isoform alpha; Protein phosphatase IA; Protein phosphatase type 1A (formely 2C) Mg-dependent alpha isoform; Protein phosphatase type 1A (formely 2C), Mg-dependent, alpha isoform; protein phosphatase, Mg2+/Mn2+ dependent 1A; protein phosphatase, Mg2+/Mn2+ dependent, 1A | |
Rabbit | |
Affinity Chromatography | |
RUO | |
19042, 24666, 5494 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
1.32 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P20650, P35813, P49443 | |
PPM1A | |
Recombinant fusion protein containing a sequence corresponding to amino acids 161-370 of human PPM1A (NP_0050291). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.