Learn More
Invitrogen™ PPM1A Recombinant Rabbit Monoclonal Antibody (006)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA529750
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Mouse PPM1A/PP2CA.
PPM1A (Protein phosphatase 1A) is also named as PPPM1A and belongs to the PP2C family. It is a serine/threonine protein phosphatase that is essential for regulating cellular stress responses in eukaryotes. PPM1A plays an important role in tumor suppression and that it negatively controls many signaling pathways which are critical for trophoblast invasion. This protein distributes in both the cytoplasm and nucleus of mammalian cells. It has 2 isoforms produced by alternative splicing.
Specifications
PPM1A | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
P20650, P35813, P49443 | |
PPM1A | |
Recombinant Mouse PPM1A/PP2CA Protein (Met1-Trp382). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Western Blot | |
006 | |
Unconjugated | |
PPM1A | |
2310003C21Rik; 2900017D14Rik; AI427932; AU017636; IA; MMPa-2; Mpp alpha; MPPa-1; Pp2c1; PP2CA; PP2Calpha; PP2C-alpha; PPM1A; PPPM1A; Protein phosphatase 1A; protein phosphatase 1A (formerly 2C), magnesium-dependent, alpha isoform; protein phosphatase 1A, magnesium dependent, alpha isoform; protein phosphatase 2C isoform alpha; Protein phosphatase IA; Protein phosphatase type 1A (formely 2C) Mg-dependent alpha isoform; Protein phosphatase type 1A (formely 2C), Mg-dependent, alpha isoform; protein phosphatase, Mg2+/Mn2+ dependent 1A; protein phosphatase, Mg2+/Mn2+ dependent, 1A | |
Rabbit | |
Protein A | |
RUO | |
19042, 24666, 5494 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.