Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PP2D1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566820
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566820 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566820 Supplier Invitrogen™ Supplier No. PA566820
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PSYQMTTDEQQ IINSFYTVFR EEYAAIEDLF SAINKTEAVR CEYEDTHKAF AKAFWRMDRL LGLGRKEVSR VQWSGC Highest antigen sequence identity to the following orthologs - mouse 72%, rat 71%.

PP2D1 gene ontology annotations related to this gene include magnesium-dependent protein serine/threonine phosphatase activity; MAP kinase serine/threonine phosphatase activity; protein serine/threonine phosphatase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PP2D1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PP2D1
Gene Accession No. A8MPX8
Gene Alias C3orf48; PP2D1; protein phosphatase 2C like domain containing 1; protein phosphatase 2C-like domain containing 1; protein phosphatase 2C-like domain-containing protein 1
Gene Symbols PP2D1
Host Species Rabbit
Immunogen Recombinant Human PP2D1. Recombinant protein control fragment (Product #RP-107476).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 151649
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.