Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PP4C Mouse anti-Human, Unconjugated, Clone: 7B5, MilliporeSigma™

Catalog No. MABE179525 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE1795-25 25 μL
MABE1795-100 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABE1795-25 Supplier MilliporeSigma Supplier No. MABE179525UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-PP4C, clone 7B5, Cat. No. MABE1795, is a highly specific mouse monoclonal antibody that targets Serine/threonine-protein phosphatase 4 catalytic subunit and has been tested for use in Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PP4C
Applications Western Blot
Classification Monoclonal
Clone 7B5
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Serine/threonine-protein phosphatase 4 catalytic subunit;EC: 3.1.3.16;Pp4;PPP4C;Protein phosphatase X;PP-X
Gene Symbols PPP4C;PPP4;PPX
Host Species Mouse
Immunogen KLH-conjugated linear peptide coreesponding to 17 amino acids from the C-terminal region of human Serine/threonine-protein phosphatase 4 catalytic subunit (PP4C).
Molecular Weight of Antigen 35.08 kDa calculated.
Purification Method Protein G purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Epigenetics & Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_002711.1
Test Specificity Clone 7B5 specifically detects human Serine/threonine-protein phosphatase 4 catalytic subunit (PP4C). It targets an epitope within 17 amino acids from the C-terminal region.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.