Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PP4R4 (PPP4R4) Monoclonal Antibody (OTI1G7), TrueMAB™, OriGene
SDP

Catalog No. 502544013
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-4013 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-4013 Supplier OriGene Supplier No. CF810585
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PP4R4 (PPP4R4) Monoclonal antibody specifically detects PP4R4 (PPP4R4) in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen PP4R4 (PPP4R4)
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI1G7
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene GCM1
Gene Accession No. Q9NP62
Gene Alias 8430415E04Rik; CFAP14; cilia and flagella associated protein 14; HEAT-like repeat-containing protein; KIAA1622; mKIAA1622; PP4R4; PPP4R4; protein phosphatase 4 regulatory subunit 4; protein phosphatase 4, regulatory subunit 4; RGD1560932; Serine/threonine-protein phosphatase 4 regulatory subunit 4
Host Species Mouse
Immunogen Full length human recombinant protein of human GCM1 produced in E.coli.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8521
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.