Learn More
PP5 Antibody (2E11), DyLight 650, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP259307C
Catalog No. NBP259307C
Only null left
Add to Cart
Description
Description
PP5 Monoclonal specifically detects PP5 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
PP5
Monoclonal
DyLight 650
50mM Sodium Borate with 0.05% Sodium Azide
PPP5C
Full length human PP5 protein
100 μL
Primary
Detects 58kDa.
Store at 4C in the dark.
IgG2a
Western Blot, Immunocytochemistry, Immunofluorescence
200000000000
Western Blot, Immunocytochemistry/Immunofluorescence
EC 3.1.3.16, PP5FLJ36922, PPP5, PPT, PP-T, protein phosphatase 5, catalytic subunit, Protein phosphatase T, Serine/Threonine phosphatase, serine/threonine-protein phosphatase 5
Mouse
Protein G purified
Cell Cycle and Replication
5536
Human
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.