Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PP5 Antibody (2E11), Novus Biologicals™

Mouse Monoclonal Antibody
$548.00
Specifications
Antigen | PP5 |
---|---|
Clone | 200000000000 |
Dilution | Western Blot 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 |
Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
Classification | Monoclonal |
Description
PP5 Monoclonal specifically detects PP5 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
PP5 | |
Western Blot 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 | |
Monoclonal | |
Purified | |
RUO | |
EC 3.1.3.16, PP5FLJ36922, PPP5, PPT, PP-T, protein phosphatase 5, catalytic subunit, Protein phosphatase T, Serine/Threonine phosphatase, serine/threonine-protein phosphatase 5 | |
PPP5C | |
IgG2a | |
Protein G purified |
200000000000 | |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Mouse | |
Cell Cycle and Replication | |
5536 | |
Full length human PP5 protein | |
Primary | |
Detects 58kDa. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title