Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PP5 Antibody (OTI2E12), Allophycocyanin/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273555APCCY7
Description
PP5 Monoclonal specifically detects PP5 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Flow Cytometry.Specifications
| PP5 | |
| Monoclonal | |
| APC-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| PPP5C | |
| Full length recombinant protein of human PP5 (NP_006238) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Canine, Primate, Monkey | |
| IgG1 |
| Flow Cytometry | |
| OTI2E12 | |
| Flow Cytometry | |
| EC 3.1.3.16, PP5FLJ36922, PPP5, PPT, PP-T, protein phosphatase 5, catalytic subunit, Protein phosphatase T, Serine/Threonine phosphatase, serine/threonine-protein phosphatase 5 | |
| Mouse | |
| Affinity Purified | |
| Cell Cycle and Replication | |
| 5536.0 | |
| Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction