Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PP5 Monoclonal Antibody (12F7), APC
GREENER_CHOICE

Catalog No. PIMA545364
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545364 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA545364 Supplier Invitrogen™ Supplier No. MA545364
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

1 μg/mL of MA5-45364 was sufficient for detection of PP5 in 15 μg of human A431 lysates by ECL analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 58kDa.

This gene encodes a serine/threonine phosphatase which is a member of the protein phosphatase catalytic subunit family. Proteins in this family participate in pathways regulated by reversible phosphorylation at serine and threonine residues; many of these pathways are involved in the regulation of cell growth and differentiation. The product of this gene has been shown to participate in signaling pathways in response to hormones or cellular stress, and elevated levels of this protein may be associated with breast cancer development. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PP5
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 12F7
Concentration 1 mg/mL
Conjugate APC
Formulation 2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.4
Gene Ppp5c
Gene Accession No. P53041
Gene Alias ANP receptor; AU020526; EGK_10788; FLJ36922; FLJ55954; OTTHUMP00000165834; PP5; PPP5; Ppp5c; PPT; PP-T; protein phosphatase 5 catalytic subunit; protein phosphatase 5, catalytic subunit; protein phosphatase T; Serine/threonine-protein phosphatase 5
Gene Symbols Ppp5c
Host Species Mouse
Immunogen Full length human PP5 protein.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5536
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.