Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PP5 Antibody (2E11), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259307G
Description
PP5 Monoclonal specifically detects PP5 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
PP5 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
PPP5C | |
Full length human PP5 protein | |
100 μL | |
Primary | |
Detects 58kDa. | |
Store at 4C in the dark. | |
IgG2a |
Western Blot, Immunocytochemistry, Immunofluorescence | |
200000000000 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
EC 3.1.3.16, PP5FLJ36922, PPP5, PPT, PP-T, protein phosphatase 5, catalytic subunit, Protein phosphatase T, Serine/Threonine phosphatase, serine/threonine-protein phosphatase 5 | |
Mouse | |
Protein G purified | |
Cell Cycle and Replication | |
5536 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction