Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PP5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588227
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588227 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588227 Supplier Invitrogen™ Supplier No. PA588227
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAMAEGERTE CAEPPRDEPP ADGALKRAEE LKTQANDYFK AKDYENAIKF YSQAIELNPS NAIYYGNRSL AYLRTECYGY ALGDATRAIE LDKKYIKGYY RRAASNMALG KFRAALRDYE TVVKVKPHDK DAKMKYQECN KIVKQKAFER AIAGDEHKRS VVDSLDIESM TIEDEYSGPK LEDGKVTISF MKELMQWYKD QKKLHRKCAY; Positive Samples: U-87MG, Hela, H460, Mouse kidney, Mouse brain, Rat testis; Cellular Location: Cytoplasm, Membrane, Nucleus.

This gene encodes a serine/threonine phosphatase which is a member of the protein phosphatase catalytic subunit family. Proteins in this family participate in pathways regulated by reversible phosphorylation at serine and threonine residues; many of these pathways are involved in the regulation of cell growth and differentiation. The product of this gene has been shown to participate in signaling pathways in response to hormones or cellular stress, and elevated levels of this protein may be associated with breast cancer development. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PP5
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.12 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Ppp5c
Gene Accession No. P53041, P53042, Q60676
Gene Alias ANP receptor; AU020526; EGK_10788; FLJ36922; FLJ55954; OTTHUMP00000165834; PP5; PPP5; Ppp5c; PPT; PP-T; protein phosphatase 5 catalytic subunit; protein phosphatase 5, catalytic subunit; protein phosphatase T; Serine/threonine-protein phosphatase 5
Gene Symbols Ppp5c
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-210 of human PPP5C (NP_0062381).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19060, 5536, 65179
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.