Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PPA2 Antibody (OTI7C1), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273563PE
Description
PPA2 Monoclonal antibody specifically detects PPA2 in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
PPA2 | |
Monoclonal | |
PE | |
PBS | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry | |
OTI7C1 | |
Western Blot, Immunohistochemistry | |
EC 3.6.1.1, FLJ20459, MGC49850, mitochondrial, pyrophosphatase (inorganic) 2 | |
Full length human recombinant protein of human PPA2(NP_008834) produced in E.coli. | |
0.1 mL | |
Cancer | |
27068 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction