Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PPA2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556817
Description
Immunogen sequence: IPLKVNSKEE NGIPMKKARN DEYENLFNMI VEIPRWTNAK MEIATKEPMN PIKQYVKDGK LRYV Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 73%.
Hydrolyzes inorganic pyrophosphate. This activity is essential for correct regulation of mitochondrial membrane potential, and mitochondrial organization and function.
Specifications
PPA2 | |
Polyclonal | |
Unconjugated | |
PPA2 | |
1110013G13Rik; FLJ20459; HSPC124; Inorganic pyrophosphatase 2, mitochondrial; Ppa2; PPase 2; pyrophosphatase (inorganic) 2; pyrophosphatase SID6-306; Pyrophosphate phospho-hydrolase 2; RGD1307965; Sid6306; SID6-306 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
27068 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H2U2 | |
PPA2 | |
Recombinant protein corresponding to Human PPA2. Recombinant protein control fragment (Product #RP-102599). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction