Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598878
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598878 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598878 Supplier Invitrogen™ Supplier No. PA598878
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PPA2 Polyclonal Antibody for Western Blot, IHC (P), ELISA

Hydrolyzes inorganic pyrophosphate. This activity is essential for correct regulation of mitochondrial membrane potential, and mitochondrial organization and function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPA2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene PPA2
Gene Accession No. Q91VM9, Q9H2U2
Gene Alias 1110013G13Rik; FLJ20459; HSPC124; Inorganic pyrophosphatase 2, mitochondrial; Ppa2; PPase 2; pyrophosphatase (inorganic) 2; pyrophosphatase SID6-306; Pyrophosphate phospho-hydrolase 2; RGD1307965; Sid6306; SID6-306
Gene Symbols PPA2
Host Species Rabbit
Immunogen Recombinant Human Inorganic pyrophosphatase 2, mitochondrial protein (180-298AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27068, 310856, 74776
Target Species Human, Mouse, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.