Learn More
PPA2 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305539AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
Hydrolyzes inorganic pyrophosphate. This activity is essential for correct regulation of mitochondrial membrane potential, and mitochondrial organization and function.Specifications
PPA2 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q9H2U2 | |
PPA2 | |
Between 284 and 334 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
PPA2 | |
FLJ20459, HSPC124, PPase 2, pyrophosphatase SID6-306, pyrophosphate phospho-hydrolase 2, SID6-306 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
27068 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.