Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPAN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106430
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106430 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106430 Supplier Invitrogen™ Supplier No. PA5106430
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PPAN.

The protein encoded by this gene is an evolutionarily conserved protein similar to yeast SSF1 as well as to the gene product of the Drosophila gene peter pan (ppan). SSF1 is known to be involved in the second step of mRNA splicing. Both SSF1 and ppan are essential for cell growth and proliferation. Exogenous expression of this gene was reported to reduce the anchorage-independent growth of some tumor cells. Read-through transcription of this gene with P2RY11/P2Y(11), an adjacent downstream gene that encodes an ATP receptor, has been found. These read-through transcripts are ubiquitously present and up-regulated during granulocyte differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPAN
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PPAN
Gene Accession No. Q91YU8, Q9NQ55
Gene Alias A230087P06Rik; Brix domain-containing protein 3; BXDC3; homolog of S. cerevisiae SSF1; Peter Pan homolog; peter pan homolog (Drosophila); Ppan; second-step splicing factor 1; SSF; Ssf1; ssf-1; SSF2; suppressor of sterile four 1; suppressor of SWI4 1 homolog
Gene Symbols PPAN
Host Species Rabbit
Immunogen A synthesized peptide derived from human PPAN(Accession Q9NQ55), corresponding to amino acid residues Q408-D458.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 235036, 298699, 56342
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.