Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PPAP2A Antibody (OTI1H4), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273564IR
Description
PPAP2A Monoclonal specifically detects PPAP2A in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
PPAP2A | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
PPAP2A | |
Full length recombinant protein of human PPAP2A (NP_003702) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI1H4 | |
Western Blot, Immunohistochemistry | |
EC 3.1.3.4, lipid phosphate phosphohydrolase 1, lipid phosphate phosphohydrolase 1a, LLP1a, LPP1PAP2a2, PAP2, PAP2a, PAP2-alpha, PAP-2aPAP2alpha2, PAPalpha1, Phosphatidate phosphohydrolase type 2a, Phosphatidic acid phosphatase 2a, phosphatidic acid phosphatase type 2A, phosphatidic acid phosphohydrolase type 2a, type 2 phosphatidic acid phosphohydrolase, type-2 phosphatidic acid phosphatase alpha | |
Mouse | |
Affinity Purified | |
Cell Cycle and Replication | |
8611.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction