Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PPAP2A Polyclonal Antibody
PPAP2A Polyclonal Antibody
Supplier: Thermo Scientific PA583881
Description
PPAP2A Polyclonal Antibody for Western Blot, IHC (P)

Specifications
PPAP2A | |
Polyclonal | |
Unconjugated | |
PLPP1 | |
35 kDa PAP; Hic53; Hpic53; hydrogen peroxide inducible protein 53; Hydrogen peroxide-inducible protein 53; Lipid phosphate phosphohydrolase 1; lipid phosphate phosphohydrolase 1a; LLP1a; Lpp1; LPP-1; magnesium-independent type 2 phosphatidic acid phosphatase; mPAP; PAP2; PAP2A; PAP-2a; PAP2a2; PAP2-alpha; PAP2alpha2; PAPalpha1; phosphatidate phosphohydrolase type 2a; Phosphatidic acid phosphatase 2a; phosphatidic acid phosphatase 2a2; phosphatidic acid phosphatase type 2A; phosphatidic acid phosphohydrolase type 2a; phospholipid phosphatase 1; Plpp1; Ppap2a; PPAP2A; LPP1; type-2 phosphatidic acid phosphatase alpha; Unknown (protein for MGC:133873); WUN | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
8611 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O14494 | |
PLPP1 | |
Recombinant protein corresponding to Human PLPP1. Recombinant protein control fragment (Product #RP-100058). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction