Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPAP2A Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986554
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6554 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6554 Supplier Bioss Supplier No. BS5423R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Phosphatidic acid phosphatase type 2 (PAP2) was originally identified as a plasma membrane enzyme that catalyses the dephosphorylation of the putative second messenger, phosphatidic acid (PA) to diacylglycerol (DG). Subsequently, multiple isoforms of PAP2 were cloned [2-5]. It was found that these enzymes dephosphorylate a number of lipid phosphates in vitro other than PA, including the potent bioactive lipids, lysophosphatidic acid (LPA) and sphingosine 1-phosphate (S1P). Therefore, they have been renamed lipid phosphate phosphatases (LPPs). Currently, there are four members of this family called LPP1, LPP1a, LPP2 and LPP3.

Specifications

Antigen PPAP2A
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene PLPP1
Gene Accession No. O14494
Gene Alias 35 kDa PAP; BOS_19759; Hic53; Hpic53; hydrogen peroxide inducible protein 53; Hydrogen peroxide-inducible protein 53; Lipid phosphate phosphohydrolase 1; lipid phosphate phosphohydrolase 1a; LLP1a; Lpp1; LPP-1; magnesium-independent type 2 phosphatidic acid phosphatase; mPAP; PAP2; PAP2A; PAP-2a; PAP2a2; PAP2-alpha; PAP2alpha2; PAPalpha1; Phosphatidate phosphohydrolase type 2a; Phosphatidic acid phosphatase 2a; phosphatidic acid phosphatase 2a2; phosphatidic acid phosphatase type 2A; phosphatidic acid phosphohydrolase type 2a; Phospholipid phosphatase 1; PLPP1; Ppap2a; type-2 phosphatidic acid phosphatase alpha; WUN
Gene Symbols PLPP1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human PPAP2A.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8611
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.