Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PPAP2A Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 179671AP150UL

Description
Phosphatidic acid phosphatase type 2 (PAP2) was originally identified as a plasma membrane enzyme that catalyses the dephosphorylation of the putative second messenger, phosphatidic acid (PA) to diacylglycerol (DG). Subsequently, multiple isoforms of PAP2 were cloned [2-5]. It was found that these enzymes dephosphorylate a number of lipid phosphates in vitro other than PA, including the potent bioactive lipids, lysophosphatidic acid (LPA) and sphingosine 1-phosphate (S1P). Therefore, they have been renamed lipid phosphate phosphatases (LPPs). Currently, there are four members of this family called LPP1, LPP1a, LPP2 and LPP3.Specifications
PPAP2A | |
Polyclonal | |
Unconjugated | |
PLPP1 | |
LLP1a, LPP1, PAP 2a, PAP2, PAP2 alpha, PAP2a, PAP2a2, PAP2alpha2, PAPalpha1, PPAP2A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
8611 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O14494 | |
PLPP1 | |
PPAP2A Fusion Protein Ag11615 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction