Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ PPAP2B (Human) Recombinant Protein

Catalog No. 89998006
Click to view available options
Quantity:
25 μg

Human PPAP2B full-length ORF ( AAH09196, 1 a.a. - 311 a.a.) recombinant protein with GST-tag at N-terminal.

  • Sequence: MQNYKYDKAIVPESKNGGSPALNNNPRRSGSKRVLLICLDLFCLFMAGLPFLIIETSTIKPYHRGFYCNDESIKYPLKTGETINDAVLCAVGIVIAILAIITGEFYRIYYLKKSRSTIQNPYVAALYKQVGCFLFGCAISQSFTDIAKVSIGRLRPHFLSVCNPDFSQINCSEGYIQNYRCRGDDSKVQEARKSFFSGHASFSMYTMLYLVLYLQARFTWRGARLLRPLLQFTLIMMAFYTGLSRVSDHKHHPSDVLAGFAQGALVACCIVFFVSDLFKTKMTLSLPAPAIRKEILSPVDIIDRNNHHNMM

Specifications

Accession Number AAH09196
Gene ID (Entrez) 8613
Name phosphatidic acid phosphatase type 2B
Preparation Method Wheat germ expression system
Quality Control Testing 125% SDS-PAGE Stained with Coomassie Blue
Quantity 25 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias Dri42, LPP3, MGC15306, PAP-2b, PAP2-b, PAP2-beta, VCIP
Gene Symbol PPAP2B
Species Wheat Germ (in vitro)
Protein Tag GST
Buffer 50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.