Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPAP2B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584909
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584909 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584909 Supplier Invitrogen™ Supplier No. PA584909
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SDLFKTKTTL SLPAPAIRKE ILSPVDIIDR NNHHNMM Highest antigen sequence indentity to the following orthologs: Mouse - 92%, Rat - 89%.

Human vascular endothelial growth factor produced in E.Coli is a double, non-glycosylated, polypeptide chain containing 165 amino acids and having a molecular mass of 38231 Dalton. The rHuVEGF is purified by proprietary chromatographic techniques.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPAP2B
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PLPP3
Gene Accession No. O14495
Gene Alias 1110003O22Rik; 2610002D05Rik; AV025606; D4Bwg0538e; D4Bwg1535e; differentially expressed in rat intestine 42; Dri42; EC 3.1.3.4; ER transmembrane protein Dri 42; lipid phosphate phosphohydrolase 3; Lpp3; MGC15306; PAP2 beta; PAP2B; PAP-2b; PAP2-b; PAP2-beta; phosphatidate phosphohydrolase type 2b; Phosphatidic acid phosphatase 2b; phosphatidic acid phosphatase type 2B; phospholipid phosphatase 3; PLPP3; Ppab2b; PPAP2B; type-2 phosphatidic acid phosphatase-beta; vascular endothelial growth factor and type I collagen inducible; Vascular endothelial growth factor and type I collagen-inducible protein; VCIP
Gene Symbols PLPP3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PLPP3. Recombinant protein control fragment (Product #RP-105572).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8613
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.