Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPAP2B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590665
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590665 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590665 Supplier Invitrogen™ Supplier No. PA590665
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: HeLa, LO2, MCF7, Mouse brain, Mouse kidnay, Rat lung, Rat brain; Cellular Location: Cell membrane, Golgi apparatus, Multi-pass membrane protein, trans-Golgi network membrane.

Human vascular endothelial growth factor produced in E.Coli is a double, non-glycosylated, polypeptide chain containing 165 amino acids and having a molecular mass of 38231 Dalton. The rHuVEGF is purified by proprietary chromatographic techniques.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPAP2B
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene PLPP3
Gene Accession No. O14495, P97544, Q99JY8
Gene Alias 1110003O22Rik; 2610002D05Rik; AV025606; D4Bwg0538e; D4Bwg1535e; differentially expressed in rat intestine 42; Dri42; EC 3.1.3.4; ER transmembrane protein Dri 42; lipid phosphate phosphohydrolase 3; Lpp3; MGC15306; PAP2 beta; PAP2B; PAP-2b; PAP2-b; PAP2-beta; phosphatidate phosphohydrolase type 2b; Phosphatidic acid phosphatase 2b; phosphatidic acid phosphatase type 2B; phospholipid phosphatase 3; PLPP3; Ppab2b; PPAP2B; type-2 phosphatidic acid phosphatase-beta; vascular endothelial growth factor and type I collagen inducible; Vascular endothelial growth factor and type I collagen-inducible protein; VCIP
Gene Symbols PLPP3
Host Species Rabbit
Immunogen Synthetic peptide.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 192270, 67916, 8613
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.