Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPAP2C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584259
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584259 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584259 Supplier Invitrogen™ Supplier No. PA584259
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PQHCLKEEEL ERKPSLSLTL TLGEADHNHY GYPHSS Highest antigen sequence indentity to the following orthologs: Mouse - 47%, Rat - 50%.

Phosphatidic acid phosphatase type 2 (PAP2) was originally identified as a plasma membrane enzyme that catalyses the dephosphorylation of the putative second messenger, phosphatidic acid (PA) to diacylglycerol (DG). Subsequently, multiple isoforms of PAP2 were cloned [2-5]. It was found that these enzymes dephosphorylate a number of lipid phosphates in vitro other than PA, including the potent bioactive lipids, lysophosphatidic acid (LPA) and sphingosine 1-phosphate (S1P). Therefore, they have been renamed lipid phosphate phosphatases (LPPs). Currently, there are four members of this family called LPP1, LPP1a, LPP2 and LPP3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPAP2C
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PLPP2
Gene Accession No. O43688
Gene Alias lipid phosphate phosphohydrolase 2; LPP2; LPR3; PAP2c; PAP-2c; PAP2-G; PAP2-gamma; phosphatidate phosphohydrolase type 2c; Phosphatidic acid phosphatase 2c; phosphatidic acid phosphatase type 2C; phosphatidic acid phosphohydrolase type 2c; phospholipid phosphatase 2; PLPP2; Ppap2c; PRG2; PRG-2; type-2 phosphatidic acid phosphatase-gamma
Gene Symbols PLPP2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PLPP2. Recombinant protein control fragment (Product #RP-91215).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8612
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.