Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PPAP2C Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA598075
Description
PPAP2C Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
Phosphatidic acid phosphatase type 2 (PAP2) was originally identified as a plasma membrane enzyme that catalyses the dephosphorylation of the putative second messenger, phosphatidic acid (PA) to diacylglycerol (DG). Subsequently, multiple isoforms of PAP2 were cloned [2-5]. It was found that these enzymes dephosphorylate a number of lipid phosphates in vitro other than PA, including the potent bioactive lipids, lysophosphatidic acid (LPA) and sphingosine 1-phosphate (S1P). Therefore, they have been renamed lipid phosphate phosphatases (LPPs). Currently, there are four members of this family called LPP1, LPP1a, LPP2 and LPP3.
Specifications
PPAP2C | |
Polyclonal | |
Unconjugated | |
PLPP2 | |
lipid phosphate phosphohydrolase 2; LPP2; LPR3; PAP2c; PAP-2c; PAP2-G; PAP2-gamma; phosphatidate phosphohydrolase type 2c; Phosphatidic acid phosphatase 2c; phosphatidic acid phosphatase type 2C; phosphatidic acid phosphohydrolase type 2c; phospholipid phosphatase 2; PLPP2; Ppap2c; PRG2; PRG-2; type-2 phosphatidic acid phosphatase-gamma | |
Rabbit | |
Protein G | |
RUO | |
8612 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
O43688 | |
PLPP2 | |
Recombinant Human Phospholipid phosphatase 2 protein (109-162AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction