Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PPAP2C Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$219.50 - $487.50
Specifications
Antigen | PPAP2C |
---|---|
Dilution | Western Blot 10 - 15 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
PPAP2C Polyclonal antibody specifically detects PPAP2C in Human samples. It is validated for Western BlotSpecifications
PPAP2C | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
EC 3.1.3.4, LPP2PAP2-g, PAP2c, PAP-2clipid phosphate phosphohydrolase 2, PAP2-G, PAP2-gamma, Phosphatidate phosphohydrolase type 2c, Phosphatidic acid phosphatase 2c, phosphatidic acid phosphatase type 2C, phosphatidic acid phosphohydrolase type 2c, type-2 phosphatidic acid phosphatase-gamma | |
Unique peptide derived from the human lysophospholipid phosphatase 2 protein. (Uniprot: O43688) | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot 10 - 15 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
PBS | |
8612 | |
IgG | |
Ammonium sulfate precipitation |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title