Learn More
PPAPDC1B Polyclonal Antibody

Description
PPAPDC1B Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | PPAPDC1B |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | PLPP5 |
| Gene Accession No. | Q3UMZ3, Q8NEB5 |
| Gene Alias | 1810019D05Rik; 2310022A04Rik; AW045471; diacylglycerol pyrophosphate like 1; diacylglycerol pyrophosphate phosphatase-like 1; DPPL1; HTPAP; phosphatidate phosphatase PPAPDC1B; phosphatidic acid phosphatase type 2 domain containing 1; phosphatidic acid phosphatase type 2 domain containing 1B; phosphatidic acid phosphatase type 2 domain-containing protein 1B; Phospholipid phosphatase 5; Plpp5; Ppapdc1; Ppapdc1b; testicular secretory protein Li 38 |
| Gene Symbols | PLPP5 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.