Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPAPDC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553722
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553722 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553722 Supplier Invitrogen™ Supplier No. PA553722
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PAHNQMDMFV TLSVDKYSFP SGHATRAALM SR Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 94%.

PPAPDC2 is a phosphatase that dephosphorylates Presqualene diphosphate (PSDP) into presqualene monophosphate (PSMP), suggesting that it may have important role in innate immunity. PSDP is a bioactive lipid that rapidly remodels to PSMP upon cell activation. PPAPDC2 displays diphosphate phosphatase activity with a substrate preference for PSDP > FDP > phosphatidic acid. PPAPDC2 activity is independent of Mg2+ and has been identified in activated human neutrophil (PMN) extracts. It is widely expressed in human tissues. Recent studies shows PPAPDC2 is a functional isoprenoid diphosphate phosphatase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPAPDC2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PLPP6
Gene Accession No. Q8IY26
Gene Alias 4932443D16Rik; bA6J24.6; FLJ46512; FLJ90191; MGC15483; PDP1; phosphatidic acid phosphatase type 2 domain containing 2; phosphatidic acid phosphatase type 2 domain-containing protein 2; Phospholipid phosphatase 6; PLPP6; polyisoprenoid diphosphate phosphatase type 1; PPAP2 domain-containing protein 2; Ppapdc2; Presqualene diphosphate phosphatase; PSDP; RP11-6J24.2
Gene Symbols PLPP6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PPAPDC2. Recombinant protein control fragment (Product #RP-91123).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 403313
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.