Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PPAR alpha/NR1C1 Antibody (3B6/PPAR), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB300537PE
Description
PPAR alpha/NR1C1 Monoclonal specifically detects PPAR alpha/NR1C1 in Human, Mouse, Rat, Bovine, Rabbit samples. It is validated for Flow Cytometry.Specifications
PPAR alpha/NR1C1 | |
Monoclonal | |
PE | |
PBS with 0.05% Sodium Azide | |
PPARA | |
Purified recombinant PPAR alpha protein. | |
0.1 mL | |
Cancer, Lipid and Metabolism, mTOR Pathway, Phospho Specific, Transcription Factors and Regulators | |
5465 | |
Human, Mouse, Rat, Bovine, Rabbit | |
Purified |
Flow Cytometry | |
3B6/PPAR | |
Flow Cytometry | |
alpha, hPPAR, NR1C1MGC2237, Nuclear receptor subfamily 1 group C member 1, peroxisome proliferator-activated receptor alpha, PPARalpha, PPAR-alpha, PPARMGC2452 | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
PPAR alpha (3B6/PPAR) | |
Store at 4C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction