Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPAR delta Polyclonal Antibody, Huabio

Catalog No. 502544681
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-4681 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-4681 Supplier HUABIO RESEARCH INC Supplier No. ER190224
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PPAR delta Polyclonal antibody specifically detects PPAR delta in Human, Mouse samples. It is validated for Flow Cytometry, Western Blot

Specifications

Antigen PPAR delta
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene Ppard
Gene Accession No. P35396, Q03181
Gene Alias FAAR; MGC3931; Nr1c2; NUC1; NUC-1; NUCI; NUCII; nuclear hormone receptor; nuclear hormone receptor 1; Nuclear receptor subfamily 1 group C member 2; OTTHUMP00000016256; OTTHUMP00000016257; peroxisome proliferative activated receptor delta; peroxisome proliferator; peroxisome proliferator activated receptor; peroxisome proliferator activated receptor beta; peroxisome proliferator activated receptor delta; peroxisome proliferator activator receptor beta; peroxisome proliferator activator receptor delta; peroxisome proliferator activator receptor, delta; peroxisome proliferator-activated nuclear receptor beta/delta variant 2; Peroxisome proliferator-activated receptor beta; Peroxisome proliferator-activated receptor delta; PPAR[b]; PPARB; Pparb/d; PPARbeta; PPAR-beta; Ppard; PPARdelta; PPAR-delta; PPARdelta/beta
Host Species Rabbit
Immunogen Recombinant protein within human PPARD aa 260-441.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19015, 5467
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.