Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPAR gamma 1 + 2Rabbit anti-Avian, Canine, Guinea Pig, Hamster, Human, Mink, Mouse, Non-human primate, Porcine, Rabbit, Rat, Rhesus Monkey, Rodent, Polyclonal, Rockland™
SDP

Catalog No. 501991734
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-1734 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-1734 Supplier Rockland Immunochemicals Supplier No. 600401419
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This affinity purified antibody is directed against human PPAR gamma protein. A BLAST analysis was used to suggest broad reactivity with this protein from several sources. Expect reactivity against human (g1 and g2), mouse (g1 and g2), rat, squirrel, orangutan, duck (g1), macaque (g1-g7), boar (g1a-1d), mink, guinea pig, rabbit (g1 and g3), dog and hamster sources based on 100% homology for the immunogen sequence. Cross reactivity with PPAR gamma homologues from other sources has not been determined. No reactivity is expected against other subtypes of PPAR.

This gene encodes a member of the peroxisome proliferator-activated receptor (PPAR) subfamily of nuclear receptors. PPARs form heterodimers with retinoid X receptors (RXRs) and these heterodimers regulate transcription of various genes. Three subtypes of PPARs are known: PPAR-alpha, PPAR-delta, and PPAR-gamma. The protein encoded by this gene is PPAR-gamma and is a regulator of adipocyte differentiation. Additionally, PPAR-gamma has been implicated in the pathology of numerous diseases including obesity, diabetes, atherosclerosis and cancer. Alternatively spliced transcript variants that encode different isoforms have been described.

Specifications

Antigen PPAR gamma 1 + 2
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene PPARG
Gene Accession No. O18924, O19052, O62807, O88275, P37231, P37238, P57797, Q4U3Q4
Gene Alias CIMT1; GLM1; HUMPPARG; Nr1c3; Nuclear receptor subfamily 1 group C member 3; OTTHUMP00000185032; OTTHUMP00000185036; peroxisome proliferator activated receptor gamma; peroxisome proliferator activated receptor gamma 2; peroxisome proliferator activated receptor gamma 4; peroxisome proliferator activator receptor, gamma; peroxisome proliferator-activated nuclear receptor gamma variant 1; Peroxisome proliferator-activated receptor gamma; PPAR gamma; PPARG; PPARG1; PPARG2; PPARgamma; PPAR-gamma; PPARgamma2; PPAR-gamma2; PPARy
Gene Symbols PPARG
Host Species Rabbit
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100008892, 100689245, 100729138, 19016, 25664, 397671, 403606, 5468, 574190
Target Species Avian, Canine, Guinea Pig, Hamster, Human, Mink, Mouse, Non-human Primate, Pig, Rabbit, Rat, Rhesus Monkey, Rodent
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.